Learn More
Medchemexpress LLC Curcumin 5-8 | 890984-26-6 | 98.58% | 339.39 | 50 MG
SDP
Catalog No. 5000362441
Encompass_Preferred
Curcumin 5-8 (CUR5-8) is a potent and orally active naturally active curcumin (CUR) analog. It inhibits lipid droplet formation, increases autophagy, and inhibits apoptosis. Furthermore, Curcumin 5-8 improves insulin resistance and insulin sensitivity.
  • Potent and orally active naturally active curcumin (CUR) analog.
  • Inhibits lipid droplet formation.
  • Increases autophagy.
  • Inhibits apoptosis.
  • Improves insulin resistance and insulin sensitivity.
Catalog No. 50-003-62441 Supplier Medchemexpress LLC Supplier No. HY14859850MG
Curcumin 5-8 (CUR5-8) is a potent and orally active naturally active curcumin (CUR) analog. It inhibits lipid droplet formation, increases autophagy, and inhibits apoptosis. Furthermore, Curcumin 5-8 improves insulin resistance and insulin sensitivity.
  • Potent and orally active naturally active curcumin (CUR) analog.
  • Inhibits lipid droplet formation.
  • Increases autophagy.
  • Inhibits apoptosis.
  • Improves insulin resistance and insulin sensitivity.