Learn More
Medchemexpress LLC Curcumin 5-8 | 890984-26-6 | 98.6% | 339.39 | 1 ML
SDP
Catalog No. 5000360628
Encompass_Preferred
Curcumin 5-8 (CUR5-8) is a potent and orally active naturally active curcumin (CUR) analog. It inhibits lipid droplet formation, increases autophagy, inhibits Apoptosis, and improves insulin resistance and insulin sensitivity.
  • Potent and orally active curcumin analog
  • Inhibits lipid droplet formation
  • Increases autophagy
  • Inhibits apoptosis
  • Improves insulin resistance and insulin sensitivity
  • Decreases palmitic acid (PA)-induced SREBP1 expression levels in AML12 cells
  • Ameliorates insulin resistance and hepatic steatosis in mice with high-fat diet-induced obesity
Catalog No. 50-003-60628 Supplier Medchemexpress LLC Supplier No. HY14859810MM/1ML
Curcumin 5-8 (CUR5-8) is a potent and orally active naturally active curcumin (CUR) analog. It inhibits lipid droplet formation, increases autophagy, inhibits Apoptosis, and improves insulin resistance and insulin sensitivity.
  • Potent and orally active curcumin analog
  • Inhibits lipid droplet formation
  • Increases autophagy
  • Inhibits apoptosis
  • Improves insulin resistance and insulin sensitivity
  • Decreases palmitic acid (PA)-induced SREBP1 expression levels in AML12 cells
  • Ameliorates insulin resistance and hepatic steatosis in mice with high-fat diet-induced obesity