Learn More
Medchemexpress LLC Curcumin 5-8 | 890984-26-6 | 98.6% | 339.39 | 25 MG
SDP
Catalog No. 5000363384
Encompass_Preferred
Curcumin 5-8 (CUR5-8) is a potent and orally active naturally active curcumin (CUR) analog. It inhibits lipid droplet formation, increases autophagy, and inhibits apoptosis. Curcumin 5-8 also improves insulin resistance and insulin sensitivity. This product is for research use only and not sold to patients.
  • Potent and orally active naturally active curcumin (CUR) analog.
  • Inhibits lipid droplet formation.
  • Increases autophagy.
  • Inhibits apoptosis.
  • Improves insulin resistance and insulin sensitivity.
  • For research use only and not sold to patients.
Catalog No. 50-003-63384 Supplier Medchemexpress LLC Supplier No. 25MGHY148598
Curcumin 5-8 (CUR5-8) is a potent and orally active naturally active curcumin (CUR) analog. It inhibits lipid droplet formation, increases autophagy, and inhibits apoptosis. Curcumin 5-8 also improves insulin resistance and insulin sensitivity. This product is for research use only and not sold to patients.
  • Potent and orally active naturally active curcumin (CUR) analog.
  • Inhibits lipid droplet formation.
  • Increases autophagy.
  • Inhibits apoptosis.
  • Improves insulin resistance and insulin sensitivity.
  • For research use only and not sold to patients.