Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Curcumin, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014222 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4222 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4222 Supplier MilliporeSigma Supelco Supplier No. 0851110MG
Only null left
Add to Cart
Add to Cart

Curcumin is an anti-inflammatory agent extracted from the roots of Curcuma longa Linn.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 458-37-7
Vapor Density 13 (vs. air)
Physical Form Powder
Linear Formula [HOC6H3(OCH3)CH=CHCO]2 CH2
MDL Number MFCD00008365
Synonym (E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione; Diferuloylmethane; Diferulylmethane; Natural Yellow 3
Type Analytical Standard
Quantity 10 mg
Formula Weight 368.38
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.