Learn More
Selleck Chemical LLC Curcumin analog C1
SDP
Catalog No. 5000002543
Encompass_Preferred
Curcumin analog C1 (Curcumin analog Compound C1) is a potent transcription factor EB (TFEB) activator with promise for the prevention or treatment of Alzheimer s disease
Catalog No. 50-000-02543 Supplier Selleck Chemical LLC Supplier No. S676910MM/1ML
Curcumin analog C1 (Curcumin analog Compound C1) is a potent transcription factor EB (TFEB) activator with promise for the prevention or treatment of Alzheimer s disease