Learn More
Medchemexpress LLC Curcumin-β-D-glucuronide | 227466-72-0 | 98.0% | 544.50 | 1 MG
SDP
Catalog No. 5000351979
Encompass_Preferred
Curcumin-β-D-glucuronide is a major metabolite found after oral intake of Curcumin in hepatic tissue and portal blood. It is utilized in research concerning colon cancer.
  • Major metabolite found after oral intake of Curcumin
  • Exhibits potent anti-tumor effects on KRASm/TP53w HCT116 xenograft model
  • Suppresses tumor growth by 38% without negatively impacting body weight
  • Attenuates NF-κB activity
  • For research use only
Catalog No. 50-003-51979 Supplier Medchemexpress LLC Supplier No. HY1387951MG
Curcumin-β-D-glucuronide is a major metabolite found after oral intake of Curcumin in hepatic tissue and portal blood. It is utilized in research concerning colon cancer.
  • Major metabolite found after oral intake of Curcumin
  • Exhibits potent anti-tumor effects on KRASm/TP53w HCT116 xenograft model
  • Suppresses tumor growth by 38% without negatively impacting body weight
  • Attenuates NF-κB activity
  • For research use only