Learn More
Cayman Chemical CurcumIn-d6 500ug
SDP
Catalog No. 502936852
Encompass_Preferred
An internal standard for the quantification of curcumin by GC- or LC-MS
Catalog No. 50-293-6852 Supplier Cayman Chemical Supplier No. 25438500 UG
An internal standard for the quantification of curcumin by GC- or LC-MS