Learn More
AdipoGen Curcumin (high purity) (10 mg)
SDP
Supplier:  AdipoGen AGCN20059M010
Encompass_Preferred
Chemical. CAS: 458-37-7. Formula: C21H20O6. MW: 368.4. Synthetic. Originally isolated from turmeric (Curcuma longa). Anti-cancer compound. Interferes with multiple cell signaling pathways, including cell cycle (cyclin D1 and cyclin E), apoptosis (activation of caspases and down-regulation of anti-apoptotic gene products), proliferation (HER-2, EGFR and AP-1), survival (PI3K/AKT pathway), invasion (MMP-9 and adhesion molecules), angiogenesis (VEGF), metastasis (CXCR-4) and inflammation (NF-kappaB, TNF, IL-6, IL-1, COX-2 and 5-LOX). Anti-angiogenic. Anti-metastatic. Anti-invasive. Chemopreventive. Sonic hedgehog (Shh) signaling pathway modulator. Downregulates Shh and Gli1. Antioxidant. Anti-inflammatory. Potent inhibitor of NF-kappaB, cyclooxygenase-2 (COX-2), lipooxygenase (LOX), and inducible nitric oxide synthase (iNOS; NOSII). Downregulates the expression of various proinflammatory cytokines including TNF, IL-1, IL-2, IL-6, IL-8, IL-12 and chemokines. Potent immunomodulator.
Catalog No. 50-205-3407