Learn More
Lkt Laboratories Curcumin, high purity, ≥98%, 25MG
SDP
Supplier:  Lkt Laboratories C807025MG
Encompass_Preferred
Active compound found in turmeric; MST1 activator, CRMP2 inhibitor. CAS: 458-37-7. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. NC1455981