Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Curcumin, Matrix Substance for MALDI-MS, ≥99.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014221 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4221 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4221 Supplier MilliporeSigma Supelco Supplier No. 78246100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 458-37-7
Vapor Density 13 (vs. air)
Physical Form Powder
Linear Formula [HOC6H3(OCH3)CH=CHCO]2 CH2
MDL Number MFCD00008365
Melting Point 183°C to 188°C
Synonym (E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione; Diferuloylmethane; Diferulylmethane; Natural Yellow 3
Type Matrix Substance for MALDI-MS
Quantity 100 mg
Formula Weight 368.38
Percent Purity ≥99.5% (HPLC)
Grade Matrix Substance for MALDI-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.