Learn More
Cayman Chemical Curcumin analog C1, Purity: ≥98%, CAS: 39777-61-2, 10 mg
SDP
Catalog No. 502949809
Encompass_Preferred
A synthetic curcuminoid and activator of TFEB; inhibits proliferation of HCT116 cells (GI50 = 4 µM); activates TFEB, inducing nuclear translocation in Neuro2a cells at 1 µM; increases LC3-II levels and promotes autophagic flux in Neuro2a cells at 1 µM; activates TFEB and increases autophagy and lysosome biogenesis in rat brain at 10 and 25 mg/kg, p.o.
Catalog No. 50-294-9809 Supplier Cayman Chemical Supplier No. 3425510 MG
A synthetic curcuminoid and activator of TFEB; inhibits proliferation of HCT116 cells (GI50 = 4 µM); activates TFEB, inducing nuclear translocation in Neuro2a cells at 1 µM; increases LC3-II levels and promotes autophagic flux in Neuro2a cells at 1 µM; activates TFEB and increases autophagy and lysosome biogenesis in rat brain at 10 and 25 mg/kg, p.o.