Learn More
Medchemexpress LLC HY-131166 500mg Medchemexpress, Curdlan CAS:54724-00-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131166500MG
Encompass_Preferred
Medchemexpress, HY-131166 500mg Curdlan CAS:54724-00-4 Curdlan is a polysaccharide produced by bacteria and a homopolymer of glucose with β-1,3-glucosidic linkage. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1266