Learn More
Apexbio Technology LLC Curdlan 54724-00-4 1g
SDP
Catalog No. 5000095920
Encompass_Preferred
Curdlan (CAS 54724-00-4) is a linear -1 3-glucan polysaccharide synthesized by bacteria It is notable for its high water solubility and ability to form thermally stable gels Mechanistically curdlan interacts with immune cell receptors such as Dectin-1 modulating cellular signaling pathways involved in innate immunity Experimental studies indicate it can stimulate macrophage activation and cytokine production suggesting immunomodulatory and potential antitumor activities In biomedical research curdlan is utilized to investigate innate immune responses as well as for its application as a gel matrix in drug delivery and tissue engineering studies
Catalog No. 50-000-95920 Supplier Apexbio Technology LLC Supplier No. A99181000
Curdlan (CAS 54724-00-4) is a linear -1 3-glucan polysaccharide synthesized by bacteria It is notable for its high water solubility and ability to form thermally stable gels Mechanistically curdlan interacts with immune cell receptors such as Dectin-1 modulating cellular signaling pathways involved in innate immunity Experimental studies indicate it can stimulate macrophage activation and cytokine production suggesting immunomodulatory and potential antitumor activities In biomedical research curdlan is utilized to investigate innate immune responses as well as for its application as a gel matrix in drug delivery and tissue engineering studies