Learn More
Med Vet International Miltex Martini Bone Curette, 5.5in., Double-Ended, 4mm And 5mm Round Cups
Catalog No. 502830076
Encompass_Preferred
The Miltex 21-321 Martini bone curette is made of OR grade German stainless steel and measures 5.5" in length. It is double-ended and the tips are 4mm and 5mm round cups.
Catalog No. 50-283-0076 Supplier Med Vet International Supplier No. MIL21321
The Miltex 21-321 Martini bone curette is made of OR grade German stainless steel and measures 5.5" in length. It is double-ended and the tips are 4mm and 5mm round cups.