Learn More
Med Vet International Miltex Williger Bone Curette, 5.5in., Double-Ended, 3mm And 4mm Oval Cups
Catalog No. 502830077
Encompass_Preferred
The Miltex 21-322 Williger bone curette is made of OR grade German stainless steel and measures 5.5" in length. It is double-ended and the tips are 3mm and 4mm oval cups.
Catalog No. 50-283-0077 Supplier Med Vet International Supplier No. MIL21322
The Miltex 21-322 Williger bone curette is made of OR grade German stainless steel and measures 5.5" in length. It is double-ended and the tips are 3mm and 4mm oval cups.