Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Curian™ Campy Test Kit
GSA_VA

Catalog No. 23315134 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-134 30 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-315-134 Supplier Meridian Bioscience™ Supplier No. 760730
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy

Curian Campy is a rapid, qualitative, fluorescent immunoassay for the direct detection of Campylobacter-specific antigen in stool specimens (for use with the Curian™ analyzer).

  • Detects C. jejuni, C. coli, C. upsaliensis, and C. lari in human stool from patients with signs and symptoms of gastroenteritis
  • Turnaround time of 20 minutes
  • Sample type includes unpreserved fecal specimens or preserved fecal specimens in transport media
  • For use with the Curian analyzer - a compact immunofluorescent instrument that boasts a simple workflow, clean sample handling, and eliminates subjectivity

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance FDA cleared
Control Sets External Controls Included
Clia Complexity Moderate
Detection Method Fluorescence Immunoassay (FIA)
Sample Type Preserved and Unpreserved Stool Specimens
For Use With (Equipment) Curian Analyzer
Includes 30 Test Device Cards, 30 Aiopreps, 33 Transfer Pipettes, Positive Control
Packaging Type Kit
Quantity 30 Tests/Kit
Species Campylobacter
Strain or Genotype C. jejuni, C. coli, C. upsaliensis, C. lari
Test Time 20 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.