Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Curian™ HpSA™ Test Kit 30T/EA
GSA_VA

Catalog No. 23315132 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-132 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-315-132 Supplier Meridian Bioscience™ Supplier No. 760130
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A rapid, qualitative, fluoresecent immunoassay for the detection of Helicobacter pylori antigens in human stool for use with the Curian™ Analyzer.

  • Intended to aid in the diagnosis of H. pylori infection and to monitor response during post-therapy
  • Lowest limit of detetion (LoD) on the market at 2 ng/
  • PPA: 96.05%; NPA: 97.00%
  • Sample type is fresh stool
  • Turnaround time of 20 minutes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance FDA cleared
Content And Storage 2°C to 8°C
CE Marker Yes
Detectable Analytes H. pylori Antigen
Type H. pylori Immunoassay
For Use With (Application) Fluorescence Immunoassay (FIA)
Includes 30 device cards, 30 Aiopreps, 32 sample brushes, wooden applicators, positive control
Packaging Type Kit
Quantity 30 Tests
Species H. pylori
Test Time 20 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.