Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience, Inc. Curian™ Shiga Toxin

Catalog No. 14910202 Shop All Meridian Bioscience Products
Change view
Click to view available options
Detectable Analytes:
E. coli Shiga Toxin 1 and 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
14-910-202 E. coli Shiga Toxin 1 and 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-202 Supplier Meridian Bioscience, Inc. Supplier No. 760630
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Curian™ Shiga Toxin is a rapid, fluorescent immunoassay for the simultaneous detection and differentiation of Shiga toxin 1 and 2 from stool specimens in broth enrichment, 30 tests/kit.

  • Fully compliant with CDC Guidelines and Joint Commission requirements for Shiga Toxin testing
  • The Curian analyzer provides a digital test result that eliminates the risk of subjective analysis by the technologist
  • All Curian assays utilize an all-in-one sample prep device, Aioprep, to allow for clean handling of stool specimens
  • The Curian Shiga Toxin assay joins Campylobacter and H. pylori to Meridian's GI focused test menu on the Curian Platform
  • Shiga Toxin 1 Sensitivity/Specificity: 100%, 99.4%
  • Shiga Toxin 2 Sensitivity/Specificity: 100%, 99.5%

Specifications

Certifications/Compliance FDA Cleared
Content And Storage 30 Tests/Kit, 2°C to 8°C
Clia Complexity Moderate
Detectable Analytes E. coli Shiga Toxin 1 and 2
Detection Method Fluorescent Immunoassay (FIA)
Format 30 Tests/Kit
Sample Type Preserved and unpreserved stool specimens
Sensitivity Shiga Toxin 1: 100%, Shiga Toxin 2: 100%
Type Fluorescent Immunoassay (FIA)
For Use With (Equipment) Curian Analyzer
Includes 30 Test Device Cards, 30 Aiopreps/Negative Control, 66 Transfer Pipettes, Positive Control
Species STEC
Specificity Shiga Toxin 1: 99.4%, Shiga Toxin 2: 99.5%
Test Time 20 min.
Analysis Time 20 min.
Dimensions (L x W x H) 9.56 x 7.25 x 4.06 in.
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.