Learn More
CURIS System CURIS 2 with Hand Sprayer
Catalog No. 01258290
Change view
Click to view available options
Product Type:
VHP Generator
Catalog No. Product Type
01-258-290 VHP Generator
1 options
Catalog No. 01-258-290 Supplier CURIS System Supplier No. CUF2IDB
Add to Cart
Add to Cart
Request a Quote
Portable CURIS 2 HHP™ Biodecontamination System with included quick connect hand sprayer provides sporicidal efficacy using EPA-registered low concentration hydrogen peroxide to disinfect/decontaminate whole rooms or targeted surfaces.
  • 36 lbs.
  • Portable Hand Carry or Trolley
  • Light-weight, ergonomic Hand Sprayer with built-in laser to easily aim and target spray surfaces.
  • Remote operation: decontaminate at the touch of a button for hands-free treatment of your entire lab.
  • Powerful ≥ 6-log efficacy using EPA-registered sporicide—validatable with biological indicators
  • Low concentration solution means less burden on staff. No monitoring required.
  • Safer for staff and surfaces than 35% hydrogen peroxide systems.
  • No harmful, sticky residues
  • Made in the USA
Specifications
Product Type VHP Generator