Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CURIS System CURoxide™ and Chemical Indicator Bundle

Catalog No. 01258291
Change view
Click to view available options
Disposable:
Single-use
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable
01-258-291 Single-use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-291 Supplier CURIS System Supplier No. COXGBCI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EPA-Registered sporicidal CURoxide™ solution (7% hydrogen peroxide) for use with CURIS 2 HHP™ Biodecontamination System and 50 pack of chemical indicator strips.

  • EPA Reg. No. 93324-1
  • Sporicidal
  • Low concentration hydrogen peroxide (7%)
  • Eliminates diff in a tri-part soil load
  • Non-Hazmat for fast, easy shipping
  • Noncorrosive composition is safer for sensitive equipment (than high-concentration solutions) and decomposes into water and oxygen
  • includes one set of 50 chemical indicators for verification the decontamination treatment has reached every corner of the space
  • Made in the USA

Specifications

Product Type Chemical Indicator Bundle
Color-coded No
Form Clear Liquid
pH 3-4
Scent Unscented
Sterility Non-sterile
Autoclavable Not Autoclavable
Disposable Single-use
Filtered Non-filtered
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.