Learn More
Lascar Electronics Inc Current Data Logger with USB
SDP
Catalog No. NC1856601
Encompass_Preferred
This standalone data logger measures more than 32,000 readings over a 4-20mA d.c. measurement range. The data logger is supplied with a cap featuring a pair of screw terminals and measurement leads terminating in crocodile clips. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery and wall mount clip.
Catalog No. NC1856601 Supplier Lascar Electronics Inc Supplier No. ELUSB4
This standalone data logger measures more than 32,000 readings over a 4-20mA d.c. measurement range. The data logger is supplied with a cap featuring a pair of screw terminals and measurement leads terminating in crocodile clips. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery and wall mount clip.