Learn More
Current Technologies Bleach-Rite™ Test Strips
Catalog No. 1441260
Change view
Click to view available options
:
4 bottles
Catalog No.
14-412-60 4 bottles
1 options
This item is not returnable. View return policy
Catalog No. 14-412-60 Supplier Current Technologies Supplier No. TS9804
This item is not returnable. View return policy
Indicate if bleach solution meets the 0.525% minimum requirement for disinfecting hard surfaces and equipment
  • Help determine if bleach disinfecting products are actually working. Bleach solutions can become non-active if the lid of a wipe canister is left open, if a bottle mixed in-house is weak / too old, or a bleach / water dual cartridge system is not spraying out a 1:10 solution
  • To use, dip one of the test strips into a sample of your bleach solution. The test strip will change color within 60 seconds; compare to the color chart on bottle. The color comparision warns if bleach solution is below the .525% (1:10 dilution) threshold for disinfection
  • Also help confirm if bleach solution is at the right concentration, so there is no need to throw it out. Don't waste time remixing if solution is strong enough
Order Info
In bottles of 50 strips
Specifications
For Use With (Application) Used to indicate if Bleach solution meets the 0.525% minimum requirement for disinfecting hard surfaces and equipment and for use on Glass, tile, laminated Plastic countertops
Test Time 60 sec.