Learn More
Fine Science Tools Friedman-Pearson Rongeurs, Stainless Steel, Curved, 14 cm
SDP
Catalog No. NC9533528
Encompass
Tip Shape: Curved/ Cup Size: 0.7mm/ Alloy / Material: Stainless Steel/ Length: 14cm/ Joint Action: Single
Catalog No. NC9533528 Supplier Fine Science Tools Supplier No. 1612114
Tip Shape: Curved/ Cup Size: 0.7mm/ Alloy / Material: Stainless Steel/ Length: 14cm/ Joint Action: Single