Learn More
MSC Unger AquaDozer Curved Floor Squeegee, 24 inch, 1 ea
Catalog No. 502868943
Encompass
This heavy-duty floor squeegee is designed for efficient liquid and debris removal in industrial and commercial settings. Its robust construction ensures durability and effective performance on various surfaces.
  • EPDM rubber blade for tough cleaning tasks and flexibility.
  • Features a 14-gauge galvanized steel frame for strength and resistance to wear and tear.
  • Curved blade design for moving water, mud, and debris.
  • Suitable for heavy-duty industrial and commercial cleaning applications.
  • Has a 24 inch blade width.
Catalog No. 50-286-8943 Supplier MSC Supplier No. 06153175
This heavy-duty floor squeegee is designed for efficient liquid and debris removal in industrial and commercial settings. Its robust construction ensures durability and effective performance on various surfaces.
  • EPDM rubber blade for tough cleaning tasks and flexibility.
  • Features a 14-gauge galvanized steel frame for strength and resistance to wear and tear.
  • Curved blade design for moving water, mud, and debris.
  • Suitable for heavy-duty industrial and commercial cleaning applications.
  • Has a 24 inch blade width.