Learn More
Fine Science Tools Standard Pattern Forceps, Stainless Steel, Curved, 14.5 cm
SDP
Supplier:  Fine Science Tools 1100114
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 3.25 x 1.6mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm
Catalog No. NC9788885