Learn More
Electron Microscopy Sciences Curved Forceps with Medium Non-serrated Tips, 152 mm,
SDP
Catalog No. 50365845
Encompass_Preferred
EMS Brand, Swiss made forceps from low carbon austenitic steel, corrosion resistance, high quality, curved tips.
Length: 152 mm
Tips: Non-serrated Tips
Catalog No. 50-365-845 Supplier Electron Microscopy Sciences Supplier No. 729106
EMS Brand, Swiss made forceps from low carbon austenitic steel, corrosion resistance, high quality, curved tips.
Length: 152 mm
Tips: Non-serrated Tips