Learn More
Electron Microscopy Sciences Curved Forceps with Medium Non-serrated Tips, 180 mm
SDP
Catalog No. NC0220045
Encompass_Preferred
EMS Brand, Swiss made forceps from low carbon austenitic steel, corrosion resistance, high quality, curved tips.
Length: 180 mm
Tips: Non-serrated Tips
This item is not returnable. View return policy
Catalog No. NC0220045 Supplier Electron Microscopy Sciences Supplier No. 729107
EMS Brand, Swiss made forceps from low carbon austenitic steel, corrosion resistance, high quality, curved tips.
Length: 180 mm
Tips: Non-serrated Tips