Learn More
Fine Science Tools Graefe Forceps, Stainless Steel, Curved, 10 cm
SDP
Supplier:  Fine Science Tools 1105410
Encompass
Tip Shape: Curved/ Tips: 1x2 Teeth/ Tip Dimensions: 0.8 x 0.7mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC9407394