Learn More
Med Vet International Curved Graft Passer, X-Large, 6.0cm dia. head
Catalog No. 5000583941
Encompass_Preferred
Used in anterior cruciate repairs such as the "over the top procedure." Using the Graft Passer lessens the risk damage to joint surfaces and ancillary structures.
Catalog No. 50-005-83941 Supplier Med Vet International Supplier No. XJOR236L
Used in anterior cruciate repairs such as the "over the top procedure." Using the Graft Passer lessens the risk damage to joint surfaces and ancillary structures.