Learn More
Med Vet International Curved Graft Passer, X-Large, 6.0cm Head, J0236L, Each
Catalog No. 5000585396
Encompass_Preferred
Used in anterior cruciate repairs such as the "over the top procedure." Using the Graft Passer lessens the risk damage to joint surfaces and ancillary structures.
Catalog No. 50-005-85396 Supplier Med Vet International Supplier No. JOR236L
Used in anterior cruciate repairs such as the "over the top procedure." Using the Graft Passer lessens the risk damage to joint surfaces and ancillary structures.