Learn More
Med Vet International Curved Graft Passer, X-Small, 2.0cm Head, J0236XS, Each
Catalog No. 5000585199
Encompass_Preferred
Used in anterior cruciate repairs such as the "over the top procedure." Using the Graft Passer lessens the risk of damage to joint surfaces and ancillary structures. Stainless steel.
Catalog No. 50-005-85199 Supplier Med Vet International Supplier No. JOR236XS
Used in anterior cruciate repairs such as the "over the top procedure." Using the Graft Passer lessens the risk of damage to joint surfaces and ancillary structures. Stainless steel.