Learn More
Fine Science Tools CURVED LEFT HANDED SCISSORS
SDP
Catalog No. NC1443948
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No
Catalog No. NC1443948 Supplier Fine Science Tools Supplier No. 1416110
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No