Learn More
Fine Science Tools Micro-Mosquito Hemostats
SDP
Catalog No. NC9263523
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1.3mm/ Clamping Length: 20mm/ Alloy / Material: Stainless Steel/ Length: 12cm
Catalog No. NC9263523 Supplier Fine Science Tools Supplier No. 1301112
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1.3mm/ Clamping Length: 20mm/ Alloy / Material: Stainless Steel/ Length: 12cm