Learn More
Electron Microscopy Sciences Curved, PTFE Coated Forceps
SDP
Catalog No. 5025216
Encompass_Preferred
Full length "PTFE" coated, 98 mm long, serrated, curved tips. No conductivity.
Catalog No. 50-252-16 Supplier Electron Microscopy Sciences Supplier No. 72976
Full length "PTFE" coated, 98 mm long, serrated, curved tips. No conductivity.