Learn More
Fine Science Tools Vannas Spring Scissors, 4mm Cutting Edge, Curved, 10.5 cm
SDP
Catalog No. NC1443467
Encompass
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 4mm/ Tip Diameter: 0.15mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No
This item is not returnable. View return policy
Catalog No. NC1443467 Supplier Fine Science Tools Supplier No. 1501910
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 4mm/ Tip Diameter: 0.15mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No