Learn More
Fine Science Tools CURVED SHARP SCISSOR
SDP
Catalog No. NC1503369
Encompass
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 6mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 5.7cm/ Serrated: No
This item is not returnable. View return policy
Catalog No. NC1503369 Supplier Fine Science Tools Supplier No. 1500001
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 6mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 5.7cm/ Serrated: No