Learn More
Fine Science Tools Fine Scissors, Tungsten Carbide, Curved, 11.5 cm
SDP
Catalog No. NC9558134
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No/ Feature: Tungsten Carbide
Catalog No. NC9558134 Supplier Fine Science Tools Supplier No. 1456912
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No/ Feature: Tungsten Carbide