Learn More
Fine Science Tools Student Vannas Spring Scissors, Curved, 9 cm
SDP
Catalog No. NC9353241
Encompass
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.35mm/ Alloy / Material: Student Stainless Steel/ Length: 9cm/ Serrated: No/ Feature: Student Quality
Catalog No. NC9353241 Supplier Fine Science Tools Supplier No. 9150109
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.35mm/ Alloy / Material: Student Stainless Steel/ Length: 9cm/ Serrated: No/ Feature: Student Quality