Learn More
AdipoGen Curvularin (1 mg)
SDP
Supplier:  AdipoGen AGCN20147M001
Encompass_Preferred
Chemical. CAS: 10140-70-2. Formula: C16H20O5. MW: 292.3. Isolated from Penicillium sp. FKI-1938 Antibiotic. Antiprotozoal. Antifungal and phytotoxic compound. Anti-inflammatory. TGF-beta signaling inhibitor Anticancer compound. Inhibits expression of inducible nitric oxide synthase (iNOS; NOSII). Cell division inhibitor. Acetylcholinesterase (AChE) inhibitor.
Catalog No. 50-205-3529