Learn More
AdipoGen Curvularin
SDP
Catalog No. 502562651
Encompass_Preferred
Chemical. CAS 10140-70-2. Formula C16H20O5. MW 292.3. Isolated from Penicillium sp. FKI-1938 Antibiotic. Antiprotozoal. Antifungal and phytotoxic compound. Anti-inflammatory. TGF-beta signaling inhibitor Anticancer compound. Inhibits expression of inducible nitric oxide synthase iNOS NOSII. Cell division inhibitor. Acetylcholinesterase AChE inhibitor.
Catalog No. 50-256-2651 Supplier AdipoGen Supplier No. AGCN20147M005
Add to Cart
Add to Cart
Chemical. CAS 10140-70-2. Formula C16H20O5. MW 292.3. Isolated from Penicillium sp. FKI-1938 Antibiotic. Antiprotozoal. Antifungal and phytotoxic compound. Anti-inflammatory. TGF-beta signaling inhibitor Anticancer compound. Inhibits expression of inducible nitric oxide synthase iNOS NOSII. Cell division inhibitor. Acetylcholinesterase AChE inhibitor.