Learn More
AdipoGen Curvulin
SDP
Supplier:  AdipoGen BVT0097M005
Encompass_Preferred
Chemical. CAS 19054-27-4. Formula C12H14O5. MW 238.2. Isolated from Curvularia sp. Phytotoxin. Microtubule assembly inhibitor. Similar to curvularin.
Catalog No. 50-256-2102