Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOTRAX™ Cushion-Ease™ Solid Modular Matting System
GREENER_CHOICE

Catalog No. 19086126 Shop All NoTrax Products
Change view
Click to view available options
Dimensions (L x W):
3 x 3 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-086-126 3 x 3 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-086-126 Supplier NOTRAX™ Supplier No. 556S0033BL
  • $149.58
    $103.65 / Each of 1

    Save $45.93

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cushion-Ease™ Solid Modular Matting System is made with a resilient rubber compound and a solid top anti-slip surface texture

  • Resilient rubber compound and unique multi-nib support design offers exceptional fatigue relief
  • Multi-nib backing for comfort and ergonomic support
  • Solid top design with an anti-slip surface texture is easy to clean
  • Easy to snap tog system for on-site custom work station platforms
  • Rubber compound is grease resistant
  • Compatible with any Cushion-Ease™ family mats
  • 551 M.D. Ramp System™ and 552 extra wide M.D.-X Ramp System™ available

Specifications

Color Black
Length (English) 3 ft.
Dimensions (L x W) 3 x 3 ft.
Thickness (English) 3/4 in.
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.