Learn More
HCL Labels Personalized Empty Label - 4" x 4" - Pack of 50
SDP
Catalog No. NC1974446
Encompass_Preferred
Hazardous Material Empty (Containment) Label. Required for marking your empty waste containers and barrels. This label should be used when no more than 1 inch of residue remains on the bottom of a drum or if the drum is empty by EPA standards; and all rings and bungs are tight. This label is customized to include your personalized information for easy use. Send in your personalized information to be pre-printed.
Catalog No. NC1974446 Supplier HCL Labels Supplier No. EM10019
Hazardous Material Empty (Containment) Label. Required for marking your empty waste containers and barrels. This label should be used when no more than 1 inch of residue remains on the bottom of a drum or if the drum is empty by EPA standards; and all rings and bungs are tight. This label is customized to include your personalized information for easy use. Send in your personalized information to be pre-printed.