Learn More
HCL Labels Personalized Hazardous Waste Label - 6" x 6" - Pack of 250
SDP
Catalog No. NC1975493
Encompass_Preferred
Hazardous Waste Containment Label. This label is compliant with both California and Federal DOT and EPA Standards regarding the labeling of hazardous waste. Adhere this label to any container or drum holding hazardous waste. This label is personalized (Custom) and can include your company name, address, phone number and any other information deemed appropriate for this specific label. Send in your personalized information to be pre-printed.
Catalog No. NC1975493 Supplier HCL Labels Supplier No. PHZW0011
Hazardous Waste Containment Label. This label is compliant with both California and Federal DOT and EPA Standards regarding the labeling of hazardous waste. Adhere this label to any container or drum holding hazardous waste. This label is personalized (Custom) and can include your company name, address, phone number and any other information deemed appropriate for this specific label. Send in your personalized information to be pre-printed.