Learn More
HCL Labels Personalized Satellite Accumulation - 6" x 6" - Pack of 250
SDP
Catalog No. NC1975524
Encompass_Preferred
This label is designed to help you comply with the labeling requirements for hazardous waste stored at or near the point of generation and defined as "satellite accumulation area" (SAA) see 40 CFR 262.34(c)(1). Send in your personalized information to be pre-printed.
Catalog No. NC1975524 Supplier HCL Labels Supplier No. HWSA0008
This label is designed to help you comply with the labeling requirements for hazardous waste stored at or near the point of generation and defined as "satellite accumulation area" (SAA) see 40 CFR 262.34(c)(1). Send in your personalized information to be pre-printed.