Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Custody Seals, 1 x 7 in.
GREENER_CHOICE

Catalog No. 05719337 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Tamper-evident Custody Seal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
05-719-337 Tamper-evident Custody Seal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-719-337 Supplier Thermo Scientific™ Supplier No. 500
  • $137.50
    $106.65 / Case of 100

    Save $30.85

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provide complete chain-of-custody and tamper evidence for samples from the point of collection to lab receipt.

Provide complete chain-of-custody and tamper evidence for samples from the point of collection to lab receipt. Thermo Scientific™ Custody Seal adhesive stickers securely seal your sample bottle and cannot be peeled off. The only way to open the sealed bottle is to break the paper label. Includes a writing area for custody signature and date for tamper-evident security. Apply at the point of sample collection, and ship samples to their destination with confidence.

  • Adhesive paper label sticks securely to glass and plastic — can't peel off
  • Writing area for custody signature and date provide chain-of-custody tracking and tamper evidence
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 1 x 7 in. Custody Seals
Width (English) 7 in.
Length (English) 1 in.
Certifications/Compliance None
Product Type Tamper-evident Custody Seal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.