Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Custom 10% NBF Prefills for DPMG Only
SDP

Catalog No. 21402244
Change view
Click to view available options
Quantity:
60 mL
180 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-402-244 60 mL
21-402-245 180 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-402-244 Supplier StatLab Supplier No. NB0230/DPMG
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Custom prefills designed for exclusive DPMG applications.

  • Containers prefilled to half capacity with 10% NBF for controlled tissue fixation.
  • 96 vials per case supporting streamlined lab throughput.
  • Configured specifically for DPMG requirements to ensure proper use.

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type 10% Neutral Buffered Formalin
Quantity 60 mL
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.