Learn More
Alkali Scientific (custom) 2D Barcode 5.0mL Cryogenic Vial, Bagged, Self Standing, External Thread, Sterile 9*9/rack, 6 racks /pk, 486 vials/Pack
SDP
Catalog No. 5000662568
Encompass_Preferred
Custom 2D barcode 5.0 mL cryogenic vial designed for secure storage and accurate tracking of biological samples in laboratory and biobank environments. Manufactured from high quality medical grade polypropylene, this vial provides excellent durability and chemical resistance at ultra low temperatures. The self standing base allows the vial to remain upright on flat surfaces, improving handling during sample preparation and storage. External thread design helps reduce the risk of contamination during opening and closing. Each vial features a clearly readable 2D barcode for reliable sample identification and automated data tracking. Sterile and supplied bagged for convenient handling. Packaged in 9×9 racks with 6 racks per pack, totaling 486 vials per pack for organized laboratory storage.
Catalog No. 50-006-62568 Supplier Alkali Scientific Supplier No. VCE6062D
Custom 2D barcode 5.0 mL cryogenic vial designed for secure storage and accurate tracking of biological samples in laboratory and biobank environments. Manufactured from high quality medical grade polypropylene, this vial provides excellent durability and chemical resistance at ultra low temperatures. The self standing base allows the vial to remain upright on flat surfaces, improving handling during sample preparation and storage. External thread design helps reduce the risk of contamination during opening and closing. Each vial features a clearly readable 2D barcode for reliable sample identification and automated data tracking. Sterile and supplied bagged for convenient handling. Packaged in 9×9 racks with 6 racks per pack, totaling 486 vials per pack for organized laboratory storage.