Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Custom 50 ppm Methyl Sulfide

Catalog No. 502401073 Shop All Phenomenex Products
Encompass_Preferred

Custom 50 ppm Methyl Sulfide 50 µg/mL in Toluene,1mL in a 2mL Ampoule

Catalog No. 50-240-1073 Supplier Phenomenex Inc Supplier No. AL0130130
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Custom 50 ppm Methyl Sulfide 50 µg/mL in Toluene,1mL in a 2mL Ampoule

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.