Learn More
StatLab Custom Amyloid 8UM Control Slides
SDP
Catalog No. 21033356
Change view
Click to view available options
Quantity:
72 Pk.
Catalog No. Quantity
21-033-356 72 Pk.
1 options
Catalog No. 21-033-356 Supplier StatLab Supplier No. CUC30040H
Add to Cart
Add to Cart
Use amyloid 8UM slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 72 Unstained Slides per pack
Specifications
Product Type Control Slide
Quantity 72 Pk.